Stampa…

Benzyl-4-hydroxybenzoate ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

A11735.09 A11735.18 A11735.36
A11735.09EA 36.8 EUR
A11735.09 A11735.18 A11735.36
Benzyl-4-hydroxybenzoate ≥99%
Benzyl-4-hydroxybenzoate

Purity: 99% CAS No.: 94-18-8 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 110-114° Beilstein Reference: 2115995

Formula: C₁₄H₁₂O₃
Punto di fusione: 110…114 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016471
CAS Number: 94-18-8
EINECS: 202-311-9

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR