Stampa…

S-Methylisothiouronium sulphate ≥98%

Fornitore: Thermo Fisher Scientific
Attenzione

A11044.0E A11044.22 A11044.30 A11044.36
A11044.0EEA 312 EUR
A11044.0E A11044.22 A11044.30 A11044.36
S-Methylisothiouronium sulphate ≥98%
S-Methylisothiouronium sulphate

Purity: 98+% CAS No.: 867-44-7 Molecular Formula: C4H12N4S2.H2SO4 Formula Weight: 278.38 Melting Point: ca 235° dec. Beilstein Reference: 3917217

Formula: C₄H₁₂N₄S₂·H₂SO₄
Punto di fusione: ∼ 235 °C (dec.)
Densità: 1,28
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013055
CAS Number: 867-44-7
EINECS: 212-759-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR