Stampa…

Aldeide benzoica ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

A10348.0B A10348.0I A10348.30
A10348.0BEA 33.3 EUR
A10348.0B A10348.0I A10348.30
Aldeide benzoica ≥99%
Aldeide benzoica

Purity: 99+% CAS No.: 100-52-7 Molecular Formula: C7H6O Formula Weight: 106.12 Flash Point: 62°(143°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.5450 Sensitivity: Air Sensitive Appearance: Colourless Odour: Almond-like Melting Point: -26° Boiling Point: 178-179° Beilstein Reference: 471223 Merck Reference: 14,1058 UN No.: 1990 MDL No.: MFCD00003299

Formula: C₆H₅CHO
Peso molecolare: 106.12 g/mol
Punto di ebollizione: 178…179 °C
Punto di fusione: –26 °C
Densità: 1.047
Flash Pt: 62 °C (143 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9 III
Merck Index: 13,01057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98811dd8cad9340872b5c4b5bfb305860eb1f81a&vid=934c1b87225e77b213e66c334d8f27ef4d924847&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR