Stampa…

N-Clorosuccinimide ≥98%

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: 1-Chloropyrrolidine-2,5-dione

A10310.30 A10310.0B A10310.18
A10310.30EA 49.4 EUR
A10310.30 A10310.0B A10310.18
N-Clorosuccinimide ≥98%
N-Clorosuccinimide

Purity: 98% CAS No.: 128-09-6 Molecular Formula: C4H4ClNO2 Formula Weight: 133.53 Physical Form: Powder Density: 1.65 Appearance: White Odour: Chlorine-like Melting Point: 146-150° Beilstein Reference: 113915 Merck Reference: 14,2164 UN No.: 3261 MDL No.: MFCD00005511


Regioselective chlorination and oxidizing reagent

Formula: C₄H₄ClNO₂
Punto di ebollizione: 216,5 °C (1013 hPa)
Punto di fusione: 146…150 °C
Densità: 1,65
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Merck Index: 14,2164

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR