Stampa…

(S)-Methyl-2-aminopropanoate hydrochloride 99%

Fornitore: Thermo Fisher Scientific

Synonyms: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

A10178.06 A10178.14 A10178.22
A10178.06EA 36.4 EUR
A10178.06 A10178.14 A10178.22
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride

Purity: 99% CAS No.: 2491-20-5 Molecular Formula: C4H9NO2.HCl Formula Weight: 139.58 Storage Temperature: 2°C to 8°C Melting Point: 106-110° Beilstein Reference: 3594033 Optical Rotation: +6.5° (c=10 in methanol) MDL No.: MFCD00063663

Formula: C₄H₉NO₂·HCl
Punto di fusione: 106…110 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00063663
CAS Number: 2491-20-5
EINECS: 219-652-4

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR