Stampa…
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Vertical
Fornitore: VWR Chemicals
99907.010EA 600 EUR
99907.010
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Vertical
Pompe Pompe a tamburo
Specially developed solvent withdrawal system for inert gas pressurizing.
  • Withdrawal system for 10 L barrels (stainless steel)
  • Perfluorelastomer o-rings
  • Includes 1 dip tube for 10 L barrels
Specially developed solvent withdrawal system for inert gas pressurizing for VWR® returnable 10 L stainless steel barrels.
Informazioni per l' accessorio: Ideal for use with flexible hose with filling nozzle (99908.001).