Stampa…

Acqua, BAKER ANALYZED®, J.T.Baker®

Fornitore: Avantor

Synonyms: Hydrogen oxide

9825.1000GLEA 47.58 EUR
9825.1000GL 9825.2500GL
Acqua, BAKER ANALYZED®, J.T.Baker®
Acqua
Formula: H₂O
Punto di ebollizione: 100 °C (1013 hPa)
Punto di fusione: 0 °C
Densità: 1 g/cm³ (4 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Visualizza i codici

Risultati del test di specificazione

Residue after Evaporation max. 5 ppm
LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
at 210 nm max. 0.005
at 254 nm max. 0.001
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 100 ng/ml
Trace Impurities (in ppb)
Aluminium (Al) max. 500
Calcium (Ca) max. 100
Iron (Fe) max. 100
Potassium (K) max. 100
Magnesium (Mg) max. 100
Sodium (Na) max. 100
Element concentrations are at time of lot release.

Specifiche

Environmentally Preferable Più sicuro dal punto di vista della salute umana e dell'ambiente
Certificazione ambientale EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR