Stampa…

Acido cloridrico 36,5 - 38,0% NF, Multi-Compendial, J.T.Baker®

Fornitore: Avantor
Pericolo

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = R

9544-02EA 111.45 EUR
9544-02 9544-05 9544-33 9544-07 9544-03 9544-08
Acido cloridrico 36,5 - 38,0% NF, Multi-Compendial, J.T.Baker®
Acido cloridrico
Formula: HCl
Punto di ebollizione: 110 °C (1013 hPa)
Punto di fusione: –30 °C
Densità: 1,18 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
Store below 25°C
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical = 0.008 %
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.008 %
NF - Bromide or Iodide Passes Test
NF - Free Bromine or Chlorine Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Sulfite (SO₃) Passes Test
NF - Assay (HCl) 36.5 - 38.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Free Chlorine (as Cl₂) ≤ 4 ppm
EP/BP - Sulfate (SO₄) ≤ 20 ppm
EP/BP - Residue on Evaporation ≤ 0.01 %
EP/BP - Assay (HCl) 35.0 - 39.0 %
JP - Assay (HCl) 35.0 - 38.0 %
JP - Identification Passes Test
JP - Residue on ignition, mg/10mL ≤ 1.0
JP - Bromide or Iodide Passes Test
JP - Bromine or Chlorine Passes Test
JP - Sulfate (SO₄) Passes Test
JP - Sulfite (SO₃) Passes Test
No Class 1, 2, 3 or other solvents are used or produced in the manufacturing or purification of the product. = 1 ppm
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report. = 5 ppm
JP - Mercury (Hg) = 0.04 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR