Stampa…

Acido formico ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™

Fornitore: Honeywell Chemicals
Pericolo

94318-250ML 94318-50ML 94318-1L
94318-250MLEA 90.83 EUR
94318-250ML 94318-50ML HONC94318-1L
Acido formico ∼98% (by titrimetric analysis) for mass spectrometry, Fluka™
Acido formico
Formula: HCO₂H
Punto di ebollizione: 100,4 °C (1.013 hPa)
Punto di fusione: 4 °C
Densità: 1,22 g/cm³ (20 °C)
Flash Pt: 49,5 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay Min. 97.5 %
Suitability for the LC-MS complying
Water (Karl Fischer) Max. 2.5 %
Non-volatile matter Max. 20 ppm
APHA Max. 15
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Barium (Ba) Max. 0.05 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.5 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.05 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 5 ppm
Sulfite (SO₃) Max. 10 ppm
Ammonium (NH₄) Max. 10 ppm
Appearance of the solution complying

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR