Stampa…

2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®

Fornitore: Merck


M-Clarity™ quality level = MQ100

93285-1G 93285-25G 93285-5G
93285-1GEA 47.7 EUR
93285-1G 93285-25G 93285-5G
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
Formula: C₁₈H₁₂N₆
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006045
CAS Number: 3682-35-7
EINECS: 222-965-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b614188da3585af8f3210f47b29434d2277b13ea&vid=31cd05efeb4d8311f54ee5242db22d2b98d9ee39&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f1bf05ec0a588c86a83713f218f64239ab9d054&vid=dbc851fb9f6226f0def1e768c5aafdd9897a6866&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78a35116ee243c85c68ff5eaf50219ad068efc50&vid=1d4e80821e795ee97e0a65cf54986f5d06218217&prid=web&clid=SAPDAM

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR