Stampa…

n-Esano 95,0, BAKER ANALYZED® HPLC, J.T.Baker®

Fornitore: Avantor

Synonyms: Hexyl hydride, Hexanen, Hexane

9304.2500EA 329.6 EUR
9304.2500 9304.1000 9304.5000
n-Esano 95,0, BAKER ANALYZED® HPLC, J.T.Baker®
n-Esano

for use in High Performance Liquid Chromatography

Formula: H₃C(CH₂)₄CH₃
Peso molecolare: 86.18 g/mol
Punto di ebollizione: 69
Punto di fusione: -94.3
Flash Pt: -22
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (by GC) min. 95 %
Residue after Evaporation max. 2 ppm
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.01 %
Ultraviolet Absorbance,(1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks).
at 210 nm max. 0.2
at 220 nm max. 0.08
at 254 nm max. 0.008
at 280 nm max. 0.005
at 350 nm max. 0.005
UV Cut-off, nm max. 192


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR