Stampa…

Acqua Cleaning, sterile

Fornitore: VWR Chemicals

Synonyms: Hydrogen oxide

911660ZTEA 177 EUR
911660ZT
Acqua Cleaning, sterile
Acqua

Sterile water for various uses in cleanrooms, such as cleaning and for diluting sterile chemicals.


  • Sterilised and packaged in a wrapped with Esteripapel® bag
  • Irradiated 8-14 kGy
  • Each bottle contains an irradiation indicator


Available as spray or liquid water.

Formula: H₂O
Peso molecolare: 18.02 g/mol
Punto di ebollizione: 100
Punto di fusione: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR