Stampa…

Safranina O soluzione

Fornitore: VWR Chemicals

911540ZAEA 69.9 EUR
911540ZA 911540ZAMP
Safranina O soluzione
Safranina O
Formula: C₂₀H₁₉ClN₄
MDL Number: MFCD00011759
CAS Number: 477-73-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR