Stampa…

Eosin-5-isothiocyanate amine-reactive fluorescent dye

Fornitore: Biotium

90091
90091.EA 242 EUR
90091.
Eosin-5-isothiocyanate amine-reactive fluorescent dye
Eosin-5-isothiocyanate

Eosin-5-isothiocyanate is a good photosensitizer and can be used to selectively label amines.


  • λEx/λ Em (pH9) = 521/544 nm
  • ε = 95,000
  • Orange red solid soluble in DMF or DMSO
  • Store at -20°C and protect from light


Eosin-5-isothiocyanate has a quantum yield of 0.57 for singlet oxygen generation, which makes it useful for photoconversion of electron-rich materials such as diaminobenzidine (DAB) or fluorescent dyes into highly electron-dense materials for high resolution electron microscopy studies. Additionally, eosin is a good fluorescence acceptor for fluorescein (FAM), dansyl and coumarin dyes in fluorescence resonance energy transfer (FRET).

Formula: C₂₁H₇Br₄NO₅S
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00082502
CAS Number: 60520-47-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR