Stampa…

Nichel (II) solfammato idrato

Fornitore: Thermo Fisher Scientific
Pericolo

89020.30 89020.18 89020.A1
89020.30EA 200 EUR
89020.30 89020.18 89020.A1
Nichel (II) solfammato idrato
Nichel (II) solfammato idrato

Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.


CAS No.: 13770-89-3 Molecular Formula: Ni(SO3NH2)2.xH2O Formula Weight: 250.86(anhy) Physical Form: Crystalline Appearance: Blue Odour: Odourless UN No.: 3077 MDL No.: MFCD00137261

Formula: H₄N₂NiO₆S₂·H₂O
Peso molecolare: 250.86 g/mol (anhy)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00137261
CAS Number: 13770-89-3
EINECS: 237-396-1
UN: 3077
ADR: 9 III

Visualizza i codici

Risultati del test di specificazione

Assay (unspecified) 15.1%-19.2% Ni

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR