Stampa…

Magnesium diboride ≥99% (metals basis)

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Magnesium boride

88149.22 88149.06 88149.14
88149.22EA 386 EUR
88149.22 88149.06 88149.14
Magnesium diboride ≥99% (metals basis)
Magnesium diboride

Purity: 99% CAS No.: 12007-25-9 Molecular Formula: B2Mg Formula Weight: 45.93 Physical Form: -100 Mesh Powder Appearance: Dark grey Odour: Odourless Melting Point: 800° dec. MDL No.: MFCD00049752

Formula: B₂Mg
Punto di fusione: 800 °C (dec.)
Densità: 2,3 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00049752
CAS Number: 12007-25-9
EINECS: 234-501-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR