Stampa…

Argento cloruro ≥95%

Fornitore: Thermo Fisher Scientific
Attenzione

87342.14 87342.22 87342.36
87342.14EA 132 EUR
87342.14 87342.22 87342.36
Argento cloruro ≥95%
Argento cloruro

Purity: 95% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: Coarse Powder Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odour: Odourless Melting Point: 455° Boiling Point: 1550° Solubility: Practically insoluble in water. Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates. Insoluble in alcohol, dilute acids Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399


Silver plating

Formula: AgCl
Peso molecolare: 143.32 g/mol
Punto di ebollizione: 1550 °C
Punto di fusione: 455
Densità: 5.56
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
ADR: 9 III
Merck Index: 13,08582

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR