Stampa…

Carbonio solfuro ≥99.9%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. per analisi

Fornitore: VWR Chemicals
Pericolo

85972.290EA 189 EUR
85972.290
Carbonio solfuro ≥99.9%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. per analisi
Carbonio solfuro
Formula: CS₂
Punto di ebollizione: 46,2 °C (1013 hPa)
Punto di fusione: –111,6 °C
Densità: 1,2632 g/cm³ (20 °C)
Flash Pt: –30 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (G.C.) min. 99,9 %
Identity (IR-spectrum) passes test
Density (20º/20º) approx. 1,26
Appearance clear and colourless
Boiling point 46 - 47 °C
Colour (Hazen) max. 10
Cadmium (Cd) max. 0,05 ppm
Calcium (Ca) max. 0,5 ppm
Chromium (Cr) max. 0,02 ppm
Cobalt (Co) max. 0,02 ppm
Copper (Cu) max. 0,02 ppm
Iron (Fe) max. 0,1 ppm
Lead (Pb) max. 0,1 ppm
Magnesium (Mg) max. 0,1 ppm
Manganese (Mn) max. 0,02 ppm
Nickel (Ni) max. 0,02 ppm
Zinc (Zn) max. 0,1 ppm
Sulfur dioxide (SO2) max. 0,00025 %
Hydrogen sulfide (as H2S) max. 0,00015 %
Benzene (G.C.) max. 0,005 %
Toluene (G.C.) max. 0,0005 %
Residue on evaporation max. 0,001 %
Water (K.F.) max. 0,01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR