Stampa…
Salviette per camera bianca, Quiltec® I
Fornitore: CONTEC
QT1-99 QT1-1212 LWQT0019 QT1-99B QT1-1212B/100 LWQT0034 QT1-66B
115-0378EA 86.4 EUR
115-0378 115-3077 115-3075 818-0301 115-3078 115-3076 115-3079
Salviette per camera bianca, Quiltec® I
Panni Panni per camera bianca Secco
Quiltec I wipes offer the highest sorptive capacity of any sealed edge wipe available. They are manufactured using two layers of knitted polyester fabric that are laminated together to form a stronger, more adsorbent wipe.
  • Most sorbent sealed edge wipe available
  • Extremely low in both particles and fibres reducing the possibility of contamination
  • Strong fabric is abrasion and chemical resistant
  • Laser-cut, which seals fibres at wipe's edge
  • Excellent sorbency with solvents
  • Laundered and packed in a Class 4 cleanroom
Additional fluid is captured withinMore Product Information