Stampa…

2-Bromoterephthalic acid, Sigma-Aldrich®

Fornitore: Merck


M-Clarity™ quality level = MQ200

8.41467.0005 8.41467.0025
8.41467.0005EA 105 EUR
8.41467.0005 8.41467.0025
2-Bromoterephthalic acid, Sigma-Aldrich®
2-Bromoterephthalic acid
Formula: C₈H₅BrO₄
Peso molecolare: 245.03 g/mol
MDL Number: MFCD00002403
CAS Number: 586-35-6
EINECS: 209-572-8

Visualizza i codici

Risultati del test di specificazione

Melting Point 300 °C
Molar Mass 245.02 g/mol
Synonyms 2-Bromo-1,4-benzenedicarboxylic acid
Hill Formula C8H5BrO4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR