Stampa…

Cyclopentylacetic acid per sintesi

Fornitore: Merck

8.41330.0010
8.41330.0010EA 0 EUR
8.41330.0010
Cyclopentylacetic acid per sintesi
Cyclopentylacetic acid
Formula: C₇H₁₂O₂
MDL Number: MFCD00001387
CAS Number: 1123-00-8
EINECS: 214-368-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR