Stampa…

Amantadine sulphate, Sigma-Aldrich®

Fornitore: Merck


M-Clarity™ quality level = MQ200

8.18383.0100
8.18383.0100EA 205 EUR
8.18383.0100
Amantadine sulphate, Sigma-Aldrich®
Amantadine sulphate
Formula: C₁₀H₁₉NO₄S
Punto di fusione: 299…301 °C
MDL Number: MFCD00077251
CAS Number: 31377-23-8
EINECS: 250-604-5

Visualizza i codici

Risultati del test di specificazione

Solubility (20 °C) insoluble
Melting Point 299 - 301 °C
Molar Mass 400.57 g/mol
pH 4.9 (100 g/l, H2O, 20 °C) (slurry)
Synonyms 1-Aminoadamantane sulfate, Adamantylamine sulfate
Hill Formula C20H36N2O4S
Chemical Formula C20H34N2 * H2SO4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR