Stampa…

D-(-)-N-Methylglucamine 99,0 resolution of racemats, zS, Sigma-Aldrich®

Fornitore: Merck

Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol

MQ200

8.18318.0250 8.18318.5000
8.18318.0250EA 60.9 EUR
8.18318.0250 8.18318.5000
D-(-)-N-Methylglucamine 99,0 resolution of racemats, zS, Sigma-Aldrich®
D-(-)-N-Methylglucamine
Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
Peso molecolare: 195.22 g/mol
Punto di fusione: 128…131 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004707
CAS Number: 6284-40-8
EINECS: 228-506-9
Merck Index: 12,06154

Visualizza i codici

Risultati del test di specificazione

Solubility 1000 g/l (25 °C)
Melting Point 128 - 131 °C
Molar Mass 195.21 g/mol
Bulk Density 300 kg/m3
Boiling Point 210 °C (1013 hPa) decomposes
pH 11 (10 g/l, H2O, 20 °C)

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR