Stampa…

Sodium hydroxymethanesulphinate dihydrate stabilizzato, SAFC®

Fornitore: MERCK PRODUCTION CHEMICALS
Attenzione

Synonyms: Sodium hydroxymethanesulphinate dihydrate(Chunks or pellets), Hydroxymethanesulphinic acid sodium salt dihydrate, Formaldehyde sodium sulphoxylate dihydrate, Sodium formaldehydesulphoxylate hydrate, Rongalite dihydrate, Sodium formaldehydesulphoxylate dihydrate

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

8.17068.1000 8.17068.9025
8.17068.1000EA 402 EUR
8.17068.1000 8.17068.9025
Sodium hydroxymethanesulphinate dihydrate stabilizzato, SAFC®
Sodium hydroxymethanesulphinate dihydrate
Formula: CH₃NaO₃S·2H₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00150598
CAS Number: 6035-47-8
EINECS: 205-739-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR