Stampa…

Glyoxylic acid 50% in soluzione acquosa, Sigma-Aldrich®

Fornitore: Merck
Pericolo


M-Clarity™ quality level = MQ200

8.14084.0250 8.14084.1000 8.14084.9010 8.14084.9025
8.14084.0250EA 27.5 EUR
8.14084.0250 8.14084.1000 8.14084.9010 8.14084.9025
Glyoxylic acid 50% in soluzione acquosa, Sigma-Aldrich®
Glyoxylic acid
Formula: C₂H₂O₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006958
CAS Number: 298-12-4
EINECS: 206-058-5
UN: 3265
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Solubility (20 °C) soluble
Boiling Point 111 °C (1013 hPa)
Vapor Pressure 10 hPa (25 °C)
Flash Point 115 °C
Refractive Index 1.4149 (20 °C, 589 nm)
Density 1.34 g/cm3 (20 °C)
pH 0.3 (H2O, 19.5 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR