Stampa…

Borane dimethyl sulphide complex

Fornitore: Merck

Synonyms: Borane-methyl sulphide complex

8.14049.0050
8.14049.0050EA 0 EUR
8.14049.0050
Borane dimethyl sulphide complex
Borane dimethyl sulphide complex
Formula: C₂H₉BS
Peso molecolare: 75.97 g/mol
Punto di fusione: –27 °C
Densità: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 2924
ADR: 3 II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR