Stampa…

3-Methylcrotonic acid

Fornitore: Merck

Synonyms: 3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid

8.06178.9025 8.06178.9050
8.06178.9025EA 0 EUR
8.06178.9025 8.06178.9050
3-Methylcrotonic acid
3-Methylcrotonic acid
Formula: C₅H₈O₂
Peso molecolare: 100.12 g/mol
MDL Number: MFCD00004366
CAS Number: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: 8 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR