Stampa…

Glyoxylic acid monohydrate, Sigma-Aldrich®

Fornitore: Merck
Pericolo


M-Clarity™ quality level = MQ200

8.04107.0025 8.04107.0100 8.04107.0250
8.04107.0025EA 40.3 EUR
8.04107.0025 8.04107.0100 8.04107.0250
Glyoxylic acid monohydrate, Sigma-Aldrich®
Glyoxylic acid monohydrate
Formula: C₂H₂O₃·1H₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149497
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Solubility (20 °C) soluble
Melting Point 46.7 °C
Molar Mass 92.05 g/mol
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR