Stampa…

EDTA sale bisodico diidrato 99.0-101.0% (by titrimetric analysis) per HPLC, Supelco®

Fornitore: Merck
Attenzione

79884-10G 79884-50G
79884-10GEA 47.3 EUR
79884-10G 79884-50G
EDTA sale bisodico diidrato 99.0-101.0% (by titrimetric analysis) per HPLC, Supelco®
EDTA sale bisodico diidrato

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Punto di fusione: 248 °C (dec., lit.)
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance (Color) Colorless or White
Appearance (Form) Powder or crystals
Titration (Kt) ZnSO4 0.1M 99.0 - 101.0 %
Suitability Test (HPLC) Corresponds to Standard
Metal Trace Analysis (ICP) Corresponds to Requirements
Aluminium (ICP) ≤ 5.0 mg/kg
Barium (ICP) ≤ 5.0 mg/kg
Bismuth (ICP) ≤ 5.0 mg/kg
Calcium (ICP) ≤ 10 mg/kg
Cadmium (ICP) ≤ 5.0 mg/kg
Cobalt (ICP) ≤ 5.0 mg/kg
Chromium (ICP) ≤ 5.0 mg/kg
Copper (ICP) ≤ 5.0 mg/kg
Iron (ICP) ≤ 5.0 mg/kg
Potassium (ICP) ≤ 50 mg/kg
Lithium (ICP) ≤ 5.0 mg/kg
Magnesium (ICP) ≤ 5.0 mg/kg
Manganese (ICP) ≤ 5.0 mg/kg
Molybdenum (ICP) ≤ 5.0 mg/kg
Nickel (ICP) ≤ 5.0 mg/kg
Lead (ICP) ≤ 5.0 mg/kg
Strontium (ICP) ≤ 5.0 mg/kg
Zinc (ICP) ≤ 5.0 mg/kg
Total Sulfur As SO4 (ICP) ≤ 50 mg/kg
Chloride (Cl) ≤ 50 mg/kg
Solubility (Method) 1.117 g in 30 ml H2O=0.1M
Appearance (Solution) Clear Colorless
Residue (Filter Test) No Residue
UV - Abs. At 260 nm ≤ 0.145
UV - Abs. At 280 nm ≤ 0.025
Recommended Retest Period 54 Months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR