Stampa…
Avantor® PUPSIT Assemblies
Fornitore: Avantor Fluid Handling
77865-528EA 0 EUR
77865-528 77865-532 77865-530
Avantor® PUPSIT Assemblies
Filtri Sistemi di filtri a maniche
Avantor's pioneering standardised filter assemblies meet PUPSIT requirements, streamline final filtration and filling, reduce wait times, and minimise operator error.
  • Standard designs save time: pre-designed, pre-configured, qualified, sterile, and ready to use filter assemblies reduce wait times significantly compared to custom solutions
  • Standard designs minimise risk: pre-designed PUPSIT assemblies are easy to use and reduce risk of operator error
  • Regulatory compliance: pre-designed solutions that comply with PUPSIT requirements outlined in the EU GMP