Stampa…
Tripod stand for flow cups
Fornitore: Industrial Physics
VF2061
769-1150EA 205.29 EUR
769-1150
Tripod stand for flow cups
Treppiedi
The spirit level is easy to balance thanks to the adjustable feet of the unit.
  • Stainless steel ring and PVC legs
  • Tripod stand with built-in spirit level
Suitable for all types of viscosity cups.