Stampa…

Sodio idrogeno carbonato 99,0-100,5 acidimetry on anhydrous substance, powder USP, FCC Haemodialysis, Macron Fine Chemicals™

Fornitore: Avantor Performance Materials

Synonyms: Sodio bicarbonato

7396-26EA 1897.54 EUR
7396-26 7396-06
Sodio idrogeno carbonato 99,0-100,5 acidimetry on anhydrous substance, powder USP, FCC Haemodialysis, Macron Fine Chemicals™
Sodio idrogeno carbonato

pH adjusters are used to adjust the pH of formulations.

Formula: NaHCO₃
Peso molecolare: 84.01 g/mol
Punto di fusione: 270 °C
Densità: 2.159 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (NaHCO₃) (dried basis)(by acidimetry) 99.0 - 100.5 %
USP - Aluminum (Al) ≤2 ppm
USP - Arsenic (As) ≤2 ppm
USP - Calcium (Ca) ≤0.01 %
USP - Carbonate (CO₃) ≤0.23 %
USP - Chloride (Cl) ≤0.015 %
USP - Copper (Cu) ≤1 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances Passes Test
USP - Iron (Fe) ≤5 ppm
USP - Limit of Ammonia ≤20 ppm
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Magnesium (Mg) ≤0.004 %
USP - Normal Carbonate Passes Test
USP - Limit of Organics ≤0.01 %
FCC - Assay (NaHCO₃) (dried basis)(by acidimetry) 99.0 - 100.5 %
FCC - Ammonia (as NH₃) Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
The net weight of the product in each container is within +/- 2% of the
netweight stated on the label.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR