Stampa…
ImpermaMat sealing mats, Axygen®
Fornitore: Corning
AM-2ML-RDIMP AM-2ML-SQIMP AM-2ML-SQIMPS AM-384-SQIMP AM-48-IMP
736-0341EA 104 EUR
736-0341 736-0347 731-0639 736-0334 736-0350
ImpermaMat sealing mats, Axygen®
Guarnizioni Sigilli per micropiastre
Axygen® ImpermaMat sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. ImpermaMats are chemically resistant, reusable sealing mats, that provide a high degree of confidence.
  • Chemical resistance - can be used in any application requiring the presence of DMSO or similar solvent
  • Contains no adhesives, no extractables to contaminate samples
  • Tight seal to the plate, no sample evaporation or well-to-well contamination
  • Usable within wide temperature range from –80 up to +121 °C
  • High contrast alphanumeric grid for