Stampa…
Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Fornitore: Thermo Fisher Scientific
156374 156372 156376 156378 156380 156382
734-3648EA 21.9 EUR
734-3648 734-3642 734-3647 734-3637 734-3646 734-3640
Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Fiasche Fiasche per colture cellulari
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
  • Consistent lot-to-lot performance
  • Sterile with SAL 10⁻²
  • Non-cytotoxic
  • Non-pyrogenic