Stampa…
VWR®, Piastre multipozzetto per colture cellulari
Fornitore: VWR Collection
734-2328EA 322 EUR
734-2328 734-2323 734-2324 734-2325 734-2326 734-2327 732-2904 732-2906 732-2907 732-2905 734-2777 734-2778 734-2779 734-2780 734-2781 734-2782 734-2798 734-2799 734-2800 734-2801 734-2802 734-2803 734-2888
VWR®, Piastre multipozzetto per colture cellulari
Micropiastre Piastre per colture cellulari
Piastre multipozzetto VWR® con trattamento al plasma in vuoto per crescita e adesione cellulare uniformi.
  • Uniform well volume ensures an equal growth surface area
  • Well surface is smooth and free from striations to maximise usable growth area
  • Raised rims on wells with uniform rings on the lid to reduce evaporation
  • Single-position lid reduces the risk of cross-contamination and handling mistakes
  • Wells are labelled with alphanumeric code for easy identification
  • Suitable for use with all common instruments and automation
  • Sterilised by electron beam irradiation