Stampa…
Provette per centrifuga, Corning®
Fornitore: Corning
430290 430291 430304 430828 430829 4558 431472
734-1827EA 233.08 EUR
734-1827 734-1828 734-1829 734-1868 734-1869 525-3414 525-0479
Provette per centrifuga, Corning®
Tubi Tubi per centrifuga
Corning 50 ml centrifuge tubes made of PP or PET.
  • Conical, with HDPE CentriStar™ or plug seal caps or without cap, sterile
  • Black printed graduations and a large white marking spot
  • Certified non pyrogenic and DNase/RNase-free
The CentriStar™ cap with leakproof design has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
Informazioni per l'ordine: Rack in schiuma e tappi CentriStar™ disponibili separatamente.
Confezione: in bulk o rack.