Stampa…
96-Well clear polystyrene microplates, Corning®
Fornitore: Corning
3366 3367 3368 3369 3370 3474 3585 3590 3591 3595 3596 3599 3628 3690 3695 3696 3697 3788 3795 3797 3798 3799 3894 3896 3897 3898 3997 9017 9018 3641 3358 3360 3361 3598 3300 7007 3841 3628BC
734-1784EA 349.99 EUR
734-1784 734-1553 734-1785 734-1786 734-1554 734-1585 734-1789 734-1790 734-1791 734-1793 734-1794 734-1796 734-1611 734-1624 734-1627 734-1628 734-1629 734-1641 734-1644 732-5543 734-1645 734-1797 734-1798 734-1657 734-1658 734-1659 734-1799 732-5567 732-5568 732-5521 734-1546 734-1548 734-1783 734-1795 734-4058 444-1020 734-2291 CORN3628BC
96-Well clear polystyrene microplates, Corning®
Micropiastre Piastre per colture cellulari
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. Cat. Nos. 3585 and 3595 have special low evaporation lids. Cat. No. 3841 is aseptically manufactured.
Visualizza i codici
Specifiche
Environmentally Preferable Produzione a basso impatto Produzione a basso impatto
Volume di lavoro 75 - 200
Certificazione ambientale RECs, ISO 14001 - Kennebunk RECs, ISO 14001 - Kennebunk