Stampa…
PCR microplates, 96-well, polypropylene, Corning® Thermowell® and Thermowell® GOLD
Fornitore: Corning
6551 3752 3753 3754
734-1766EA 196.4 EUR
734-1766 732-5590 732-5591 732-5592
PCR microplates, 96-well, polypropylene, Corning® Thermowell® and Thermowell® GOLD
Micropiastre Piastre per PCR
PP, non sterile. Compatible with a full range of PCR, sequencing and real-time PCR equipment.
Thermowell® GOLD 96-well PCR plates are available in different formats to ensure maximum flexibility and a perfect match for specific applications. The original Thermowell® 96-well PCR plates are universal fit and can be cut into 3×8-well segments.
Polyethylene and aluminium sealing tapes provide optimal sealing to prevent evaporation and allow for oil-free operation when used with thermal cyclers with heated lids. Silicone rubber sealing mats are fully autoclavable and reusable. The 8-wellMore Product Information