Stampa…
Filter plates, 96-well, Corning® FiltrEX™
Fornitore: Corning
3504 3508 3510 3511 3505
732-5509EA 1696.64 EUR
732-5509 732-5511 732-5512 732-5513 732-5510
Filter plates, 96-well, Corning® FiltrEX™
Micropiastre Piastre filtranti
FiltrEX™ filter plates meet the industry standards for plate dimensions. The rigid side walls make the plate ideal for automation and the wide skirt accepts barcodes. Individual filter discs are encapsulated in the plate by a process that ensures 100% integrity of each well. The design of the nozzle prevents sample wicking and cross-contamination. Glass fibre filter plates can be used for a variety of applications, such as plasmid isolation, DNA purification, PCR clean-up or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane can be used for lysate clarification,More Product Information
Visualizza i codici
Specifiche
Environmentally Preferable Produzione a basso impatto
Certificazione ambientale RECs, ISO 14001 - Kennebunk