Stampa…
TapeStation System, 4200
Fornitore: AGILENT
G2991BA
732-3992EA 0 EUR
732-3992
TapeStation System, 4200
Sistemi per elettroforesi Sistemi per elettroforesi capillare
DNA and RNA sample quality control with flexible throughput (1 to 96) at a constant cost per sample.
  • Simplify your workflow with fully automated sample processing and ready-to-use ScreenTape technology
  • Any sample number between 1 and 96 can be analysed, with constant costs per sample providing scalable throughput
  • Easily switch between DNA and RNA ScreenTape assays for excellent flexibility
  • Results are obtained quickly in as little as 1 to 2 minutes per sample, or less than 90 minutes for 96 samples
  • Achieve user-independent results with minimal manual