Stampa…
Filter tips, PIPETMAN® Diamond
Fornitore: GILSON
F171403 F171103 F171203 F171803 F171503 F171603 F171703 F171303 F172303 F172703 F172403 F172203 F172503 F172603 F172313 F172513 F172613 F172213 F172803
732-3402EA 145 EUR
732-3402 732-3399 732-3400 732-3406 732-3403 732-3404 732-3405 732-3401 732-3952 732-3967 732-3958 732-3946 732-3961 732-3965 732-3955 732-3964 732-3966 732-3949 732-3856
Filter tips, PIPETMAN® Diamond
Puntali per pipette
PIPETMAN® DIAMOND Sterilized Filter Tips are designed to fit your PIPETMAN perfectly and play a critical role in preventing aerosol-born contamination. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
  • Accurate and precise
  • Reliable
  • Convenient packaging
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing.More Product Information