Stampa…
VWR®, Cap Mat Applicator
Fornitore: VWR Collection
732-3335EA 2070 EUR
732-3335
VWR®, Cap Mat Applicator
Applicatori per film Applicatori di pellicola sigillante
Compact and portable, the cap mat applicator is very easy to use, requiring only one operation of the system to produce an accurate and tight seal on a wide range of both collection and shallow well microplates.
  • Needs minimal pressure for capping, reducing fatigue and RSI
  • Powder coated to resist chemical spillage