Stampa…
PCR detection plates, 96-well, semi-skirted, flat deck, Thermo-Fast®
Fornitore: Thermo Fisher Scientific
AB-1400 AB-1400/W AB-1400/B AB-1400/G AB-1400/P AB-1400/R AB-1400/Y AB-1400/K AB-1400-L AB-1400/B-L AB-1400/G-L AB-1400/Y-L AB-1400/W-L AB-1400/R-L AB-1400/P-L AB-1400/150
732-0892EA 162 EUR
732-0892 732-0894 731-0295 731-0297 731-0299 731-0300 731-0301 732-0893 732-0882 731-0296 731-0298 732-2953 732-2952 732-2951 732-2950 ADVAAB-1400/150
PCR detection plates, 96-well, semi-skirted, flat deck, Thermo-Fast®
Micropiastre Piastre per PCR
PP. The PCR detection plate features a flat deck for improved handling and sealing.
  • Directly compatible with all major thermal cyclers, including all ABI thermal cyclers, qPCR instruments and sequencers
  • Flat deck for improved sealing characteristics - can be sealed with heat seals, adhesive seals, mats or cap strips
  • Cut corner (A12)
  • Certified free from DNase, RNase and human genomic DNA