Stampa…
Sealing mats for 1,0 ml DeepWell™ plates, Nunc™
Fornitore: Thermo Fisher Scientific
278616 278617
732-2676EA 238 EUR
732-2676 732-2677
Sealing mats for 1,0 ml DeepWell™ plates, Nunc™
Guarnizioni Sigilli per micropiastre
EVA. For sealing PP and PS plates.
  • Efficient seal with minimum protrusion into each well
  • Prevents evaporation and dust