Stampa…
Sealing film, self-adhesive for ELISA, Real-Time PCR (qPCR)
Fornitore: Brand
781391
732-1475EA 225 EUR
732-1475
Sealing film, self-adhesive for ELISA, Real-Time PCR (qPCR)
Guarnizioni Sigilli per micropiastre
Highly transparent film made of polyester is suitable for Real-Time PCR measurements. Temperature range: −40 to +120 °C.