Stampa…
PCR tubes, 0,2 ml, and cap strips, Axygen®
Fornitore: Corning
PCR-0208-A PCR-0208-G PCR-0208-Y PCR-02CP-A PCR-02CP-C PCR-2CP-RT-C PCR-02-FCP-C PCR-12-FCP-C
731-4006EA 135.28 EUR
731-4006 731-4008 731-4012 731-4013 732-0680 732-0681 732-1375 731-0642
PCR tubes, 0,2 ml, and cap strips, Axygen®
Tubi Tubi per PCR
Axygen® polypropylene 8/12-tube thin wall PCR tube strips and caps are specifically designed for PCR use and are ideal for thermal transfer applications. The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Made from highly polished moulds and free of lubricants, dyes, heavy metals, and fillers. Certified RNase, DNase, and pyrogen free to ensure the purity of your reaction.
  • Pareti uniformi particolarmente sottili per la