Stampa…
Microarray plates, HTA™ Plate
Fornitore: Greiner Bio-One
791830_5 791840_5 791850_5
731-3018EA 1770 EUR
731-3018 731-3019 731-3020
Microarray plates, HTA™ Plate
Micropiastre Piastre microarray
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA™ Plate has been optimised for the simultaneous analysis of multiple samples.
  • Available with multifunctional, 3-D Amino, 3-D Aldehyde or 3-D Epoxy surface functionality
  • Partition of the plate into four individual strips containing 24 wells each gives flexibility, permitting processing of a variable number of samples each day
  • Plate dimensions are compatible with automated systems
  • Low well rim of only 0,3 mm
  • Low autofluorescence