Stampa…
Blood collection tubes with gel and lithium heparin anticoagulant, vacuum, Vacutest®
Fornitore: KIMA SAS DI CHIARIN RENZO
12010 12030 12570 12550 12020 12057 12005
720-2737EA 53.1 EUR
720-2737 KIMA12030 720-2740 720-2739 KIMA12020 720-2764 KIMA12005
Blood collection tubes with gel and lithium heparin anticoagulant, vacuum, Vacutest®
Sistemi di diagnostica Raccolta/Preparazione di Campioni Clinici Blood Sample Collection/Preparation
PET, transparent.
  • Available in three sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
  • 18 months shelf-life
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use,More Product Information