Stampa…
Blood collection tubes with sodium heparin, vacuum, Vacutest®
Fornitore: KIMA SAS DI CHIARIN RENZO
123100 123300 12340
720-2690EA 356 EUR
720-2690 720-2691 720-2692
Blood collection tubes with sodium heparin, vacuum, Vacutest®
Sistemi di diagnostica Raccolta/Preparazione di Campioni Clinici Blood Sample Collection/Preparation
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental fails. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in three sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
  • 18