Stampa…
UDK Kjeldahl distillation units
Fornitore: VELP SCIENTIFIC
F30200145 F30200125 F30200135 F30200155 F30200165 S30200165
701-0296EA 9130 EUR
701-0296 701-0294 701-0295 701-0297 701-0298 701-0299
UDK Kjeldahl distillation units
Sistemi di distillazione Sistemi di distillazione Kjeldahl
The UDK Series distillers are designed to meet the most challenging demands and requirements for diverse applications, according to international standards: Kjeldahl nitrogen TKN, proteins, ammoniacal nitrogen, nitric nitrogen (Devarda), phenols, TVBN and volatile acids, cyanides, and alcohol content. Five different UDK models are available with different automation levels to match any laboratory requirement of automation and throughput. A complete range of distillers featuring exclusive technologies to meet any laboratory requirement for the determination of analytes in different fields ofMore Product Information